Omnifit® EZ Chromatography Columns with Two Fixed Endpieces, Diba Industries
Supplier: Diba Industries
Omnifit®
11943-01 11943-37 11943-46 11942-86 11943-10 11942-80 11943-52 11943-43 11943-40 11943-04 11942-74 11943-58 11943-16 11943-79 11942-83 11943-73 11943-70 11943-31 11942-77 11942-92 11943-76 11943-07 11943-28 11943-25 11943-55 11943-61 11943-22 11942-95 11943-19 11942-98 11943-85 11942-89 11943-13 11943-49 11943-34 11943-82 11943-67 11943-64
76176-080EA 885.63 CAD
76176-080 76175-836 76175-758 76175-812 76175-734 76175-854 76175-776 76175-830 76175-752 76175-794 76175-770 76176-106 76176-068 76176-244 76176-062 76175-806 76175-728 76176-086 76176-092 76175-848 76175-824 76176-050 76175-746 76175-800 76175-788 76175-842 76175-764 76175-740 76175-860 76175-980 76175-782 76176-238 76176-232 76176-056 76176-074 76176-250 76175-818 76176-098
Omnifit® EZ Chromatography Columns with Two Fixed Endpieces, Diba Industries
Chromatography Columns
PEEK™ column threads offer outstanding durability.
  • Precision borosilicate glass columns provide inertness, visibility, and performance
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Delivery information: Delivered with two (0.5 m and 2 m length) ¹/₆" Int.Ø PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 µm PE, 10 µm PTFE, and 30 µm PTFE frits), and a Click-N-Seal® 10-32 PEEK conical port fitting.