Separating Funnels, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
298280-0010 298280-0025 298281-0010 298281-0025
KT298280-0010EA 423.31 CAD
KT298280-0010 KT298280-0025 KT298280-0125 KT298281-0010 KT298281-0025
Separating Funnels, DWK Life Sciences
Funnels Separating Funnels
Micro separating funnels manufactured from 33 expansion, low extractable borosilicate glass
Squibb-type funnel with a PTFE stopcock plug with a 2 mm bore.
Certifications: Material conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery information: Series 298280 is supplied with hex-head stopper and series 298281 is supplied without stoppers.