Avantor® Genesis, C8 HPLC Columns
Supplier: Avantor
FK5962E FM5964 FM10962E FM15962E FM25962E FH1962-2/N FM25964E GEN-4C8-125DF GEN-5C8-150D GEN-5C8-250D
75798-684EA 2597.13 CAD
75798-684 75798-994 75798-996 75798-998 75799-000 75799-002 75799-004 75830-394 75830-608 76398-868 76398-872 76398-874 76398-880 76398-902 76398-904 76398-922 76398-924
Avantor® Genesis, C8 HPLC Columns
Chromatography Columns
Avantor® Genesis phases are based on high purity metal-free spherical silica.
  • Good peak shape and reproducibility
  • Long column lifetime
  • pH stability 1 to 10
  • Stainless steel column hardware
They are suitable for the analysis of a wide range of analytes.