WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
11-0051G 11-0051K 11-0051R 11-0053DB 11-0053K 11-0053N 11-0054DB 11-0054G 11-0054GT 11-0054N 11-0054NT 11-0054R 11-0054RT 11-0055G 11-0055GT 11-0055K 11-0055R 11-0051DB-B 11-0051G-B 11-0051K-B 11-0051N-B 11-0051R-B 11-0053DB-B 11-0053G-B 11-0053K-B 11-0053N-B 11-0053R-B 11-0054DB-B 11-0054G-B 11-0054K-B 11-0054N-B 11-0054R-B 11-0055DB-B 11-0055R-B 11-0056DB 11-0056K 11-0056N 11-0056R 242772-06 242776-01 242776-02 242776-04 242776-05 242772-01 242772-02 242772-04 242772-05
97047-114PK 60.48 CAD
97047-114 97047-116 97047-118 97047-120 97047-122 97047-124 97047-126 97047-128 97047-130 97047-132 97047-134 97047-136 97047-138 97047-140 97047-142 97047-144 97047-146 97047-148 97047-150 97047-152 97047-154 97047-156 97047-158 97047-160 97047-162 97047-164 97047-166 97047-168 97047-170 97047-172 97047-174 97047-176 97047-178 97047-180 97047-182 97047-184 97047-186 97047-188 97047-190 97047-192 97047-194 97047-196 97047-198 97047-200 97047-202 97047-204 97047-206 97047-208 97047-210 97047-212 97047-214 97047-216 97047-218 97047-220 97047-222 97047-224 97047-226 97047-228 97047-230 97047-232 97050-382 97050-383 97050-384 97050-385 97050-386 97050-387 97050-388 97050-389 97050-390 97050-391 97050-392 97050-393 97050-394 97050-395 97050-396 97050-397 97050-398 97050-399 97050-400 97050-401 97050-402 97050-403 97050-404 97050-405 97050-406 97050-407 97050-408 97050-409 97050-410 97050-411 97050-412 97050-413 97050-414 97050-415 97050-416 97050-417 66014-630 89235-558 89235-560 89235-562 89235-564 89235-568 89235-570 89235-572 89235-574
WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
Closures Chromatography Vial Closures
These snap-top caps eliminate the need for crimping and decapping tools.
  • Manufactured from PE
  • Fits the WHEATON E-Z Vials® with snap ring top
Available in multiple colors and septa types.
Order Now
SPECIFICATIONS
Environmentally Preferable