Formvar Film on Thin Bar Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FFT200-CU-50 FFT200-CU-UL FFT300-CU-UL FFT400-CU-UL FFT1000-CU-UL FFT200-NI-TH FFT300-NI-TH FFT400-NI-TH FFT200-CU-TH FFT300-CU-TH FFT400-CU-TH FFT1000-CU-TH FFT200-CU-ET FFT300-CU-ET FFT400-CU-ET FFT1000-CU-ET FFT200-NI-UL FFT300-NI-UL FFT400-NI-UL FFT1000-NI-UL FFT1000-NI-TH FFT200-NI-ET FFT300-NI-ET FFT400-NI-ET FFT1000-NI-ET FFT200-AU-UL FFT300-AU-UL FFT400-AU-UL FFT200-AU-ET FFT300-AU-ET FFT400-AU-ET FFT200-AU-TH FFT300-AU-TH FFT400-AU-TH
102100-988BX 228.25 CAD
102100-988 103303-830 103303-832 103303-834 103303-836 103303-972 103303-974 103303-976 103304-064 103304-066 103304-068 103304-070 103304-276 103304-278 103304-280 103304-282 103304-988 103304-990 103304-992 103304-994 103305-082 103305-124 103305-126 103305-128 103305-130 103305-584 103305-586 103305-588 103305-618 103305-620 103305-622 103306-034 103306-036 103306-038
Formvar Film on Thin Bar Square Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a Thin Bar Square grid. Coated grids are optically checked and batch tested in the electron microscope.
  • Select from copper, nickel, and gold grids in 200, 300, 400 or 1000 mesh sizes
  • Optically checked followed by batch testing in the EM
Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm).
Packaging: Packed in grid storage box.