Omnifit® EZ Chromatography Columns with Two Adjustable Endpieces, Diba Industries
Supplier: Antyila Scientific
11942-90 11942-93 11942-96 11942-99 11943-05 11943-08 11943-11 11943-14 11943-17 11943-20 11943-23 11943-26 11943-53 11943-56 11943-59 11943-62 11943-65 11943-68 11943-71 11943-29 11943-32 11943-35 11943-38 11943-41 11943-44 11943-47 11943-50 11943-74 11943-77 11943-80 11943-83
76175-748EA 1178.84 CAD
76175-748 76175-754 76175-760 76175-766 76175-778 76175-784 76175-790 76175-796 76175-802 76175-808 76175-814 76175-820 76175-826 76175-832 76175-838 76175-844 76175-850 76175-856 76175-862 76176-058 76176-064 76176-070 76176-076 76176-082 76176-088 76176-094 76176-100 76176-108 76176-234 76176-240 76176-246
Omnifit® EZ Chromatography Columns with Two Adjustable Endpieces, Diba Industries
Chromatography Columns
Ergonomically designed endpieces provide simple and quick height adjustment.
  • PEEK™ column threads offer outstanding durability
  • Precision borosilicate glass columns provide inertness, visibility, and performance
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Delivery information: Delivered with two (0.5 and 2 m length) ¹/₁₆" Ext.Ø PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 µm PE, 10 µm PTFE, and 30More Product Information