EZ-Decappers™ Hand Operated Cappers and Decappers, MicroLiter
Supplier: DWK Life Sciences (KIMBLE)
08-0000-CEZ 11-0000-CEZ 13-0000-CEZ 20-0000-CEZ 08-0000-DEZ
89426-106EA 812.27 CAD
89426-106 89426-108 89426-110 89426-112 89426-114
EZ-Decappers™ Hand Operated Cappers and Decappers, MicroLiter
Crimpers, Cappers and Decappers
EZ-Decappers™ Hand Operated Cappers and Decappers by MicroLiter are easy-to-use tools for crimping and decrimping chromatography vial closures.
  • Easy-to-use
  • Multiple seal sizes available