Vydac 214TP C4, HPLC Columns
Supplier: Avantor
214TP5205 214TP5405 214TP5210 214TP5410 214TP5115 214TP5215 214TP5315 214TP5415 214TP51 214TP52 214TP53 214TP54 214TP10415 214TP104 214TP1022 214TP10154 214TP510 214TP101015 214TP1010 214GCC1210 214FSK1210 214FSK1510 214TP10152215 214TP101510 214FSK510 214GCC510 214TP101522 214GCC152010 214TP152022 214GD104/N 214GD52/N 214GD54/N 214GK104/N 214GK52/N 214GK54/N 214GCC1010 214GCC1520/N 214GD51/N 214GK51/N 5138110
75798-390EA 2027.95 CAD
75798-390 75798-392 75798-394 75798-398 75798-400 75798-402 75798-404 75798-406 75798-410 75798-412 75798-414 75798-418 75798-420 75798-738 75798-740 75798-742 75798-744 75798-746 75798-750 75798-752 75798-756 75798-762 75798-764 75798-766 75798-768 75798-770 75812-876 75812-878 75812-882 75812-884 75812-886 75812-888 75812-890 75812-892 75812-894 75812-896 75812-898 75813-014 75813-016 75813-018 75813-020 75813-022 75813-024 75813-026 75813-028 75830-750 75830-752 75830-756 75830-760 75830-766 75830-768 75830-772 75830-776 76398-046 76398-048 76398-050 76398-052 76398-056 76398-062 76398-334
Vydac 214TP C4, HPLC Columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L26
214TP C4 is a first generation C4 phase and is recommended for the analysis of glycoproteins, haemoglobin variants, histones, insulin variants and membrane proteins.
The large pores of the 300 Å TP silicaMore Product Information