Vydac 214ATP C4, HPLC Columns
Supplier: Avantor
214ATP5405 214ATP54 214ATP5415 214AGD54/N
75798-396EA 1699.2 CAD
75798-396 75798-408 75798-416 75798-736 75798-748 75798-754 75812-880 75830-582 75830-584 75830-748
Vydac 214ATP C4, HPLC Columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L26
214ATP C4 has a lower level of endcapping and is optimized for the analysis of human growth hormone.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.