Formvar Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FFTH200-CU-25 FFTH400-CU-50 FFTH200-CU-UL FFTH300-CU-UL FFTH400-CU-UL FFTH600-CU-UL FFTH200-CU-TH FFTH300-CU-TH FFTH400-CU-TH FFTH600-CU-TH FFTH200-CU-ET FFTH300-CU-ET FFTH400-CU-ET FFTH600-CU-ET FFTH200-NI-UL FFTH300-NI-UL FFTH400-NI-UL FFTH600-NI-UL FFTH200-NI-TH FFTH300-NI-TH FFTH400-NI-TH FFTH600-NI-TH FFTH200-AU-UL FFTH200-AU-TH FFTH300-AU-TH FFTH400-AU-TH FFTH600-AU-TH FFTH200-AU-ET FFTH300-AU-ET FFTH400-AU-ET FFTH600-AU-ET FFTH200-NI-ET FFTH300-NI-ET FFTH400-NI-ET FFTH600-NI-ET FFTH300-AU-UL FFTH400-AU-UL FFTH600-AU-UL
102101-022BX 178.54 CAD
102101-022 102101-048 103303-838 103303-840 103303-842 103303-844 103304-072 103304-074 103304-076 103304-078 103304-366 103304-368 103304-370 103304-372 103305-032 103305-034 103305-036 103305-038 103305-092 103305-094 103305-096 103305-098 103305-592 103305-604 103305-606 103305-608 103305-610 103305-702 103305-704 103305-706 103305-708 103305-918 103305-920 103305-922 103305-924 103306-000 103306-002 103306-004
Formvar Film on Thin Bar Hexagonal Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure formvar resin coats the shiny side of a thin bar hexagonal grid.
  • Select from copper, nickel, and gold grids in 200, 300, 400, or 600 mesh sizes
  • Optically checked, followed by batch testing in the EM
Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm).
Packaging: Packed in grid storage box.