Graticules with Cross Scales, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67994-21 67989-16 67989-21 67989-19 67994-16 67994-19 67978-21 67978-23 67978-24 67978-26 67978-27 67978-50 67979-23 67979-24 67979-26 67979-27 67979-50 67989-20.4 67989-23 67989-24.5 67989-25 67989-26 67989-27
100499-078EA 335.39 CAD
100499-078 100499-050 100499-054 100499-052 100499-074 100499-076 102096-020 102096-022 102096-024 102096-026 102096-028 102096-030 102096-032 102096-034 102096-036 102096-038 102096-040 103305-404 103305-406 103305-408 103305-410 103305-412 103305-414
Graticules with Cross Scales, Electron Microscopy Sciences
Graticules, Reticles and Grids
The graticule is used as a a horizontal and a vertical scale and is especially useful for measuments also in in different axis.
  • Scales in mm and inches available
  • Use on variety of different shaped objects
  • Extended size range