VWR® High Performance Single Channel Pipettors and Pipettor Kits
Supplier: VWR
VWR®
FF67003 FF67002 FF10001 FF10008 FF67004 FF10004 FF10003 FF10006 76208-510 76208-488 FF10005 FF10007 FF10002 FF67001
76169-240EA 280.89 CAD
76169-240 76169-262 76169-274 76169-266 76169-244 76169-234 76169-264 76169-242 76169-232 76169-238 76169-268 76169-236 76208-510 76208-488
VWR® High Performance Single Channel Pipettors and Pipettor Kits
Pipettes Micropipettes
VWR® High Performance Pipettors are the newest lightweight pipettor designed for user comfort throughout daily work. Thanks to low pipetting forces, users get a unique pipetting experience. With its high accuracy and precision, the VWR® High Performance Pipettor delivers highly precise and reproducible results.
  • Exclusive volume lock mechanism
  • Lightweight design
  • Low pipetting forces
  • Outstanding precision and accuracy
  • Fully autoclavable without disassembly (120 °C/0.1 MPa/20 min.) and UV resistant
  • Universal tip fitting
  • Unique serial number
VWR® High Performance Pipettors have been designed to fit both right- and left-handed users while providing maximum comfort.
Pipettors are available in a large volume range, with 8 single-channel designs from 0.2 µl to 10 ml. They can be equipped either with plastic or stainless steel tip ejectors (5 mL and 10 mL single-channel devices do not have a tip ejector).
The large ejector button is built to fit either hand perfectly and allows pipetting without fatigue. A single button is used to eject and secure the volume setting. A unique instant volume locking system mechanism prevents accidental volume change while in use. These features combine to allow easy one-handed operation. Simply pull the ejection button, adjust the volume with the push button, and lock the pipettor by pushing down the ejection button. Digits are easy to read and adjust. The volume is always visible while pipetting. Easy to maintain, VWR® High Performance Pipettors are developed to simplify daily use thanks to intuitive in-lab disassembly, maintenance, and a high chemical compatibility.
Certifications: CE marked.
Ordering information: Each kit contains the pipettor volumes listed in the table below, a 7-place carrousel stand, and VWR® pipette tips. Each order also includes a 36 month warranty. The product comes with a certificate of conformity, a calibration key, a Quick start guide and a tip ejector extension (for 2 and 10 µl model only).