Burned Fireclay Super Scorifiers, HarbisonWalker
Supplier: HarbisonWalker
124371 124372 141591
80701-894CS 1930.74 CAD
80701-894 11019-271 76018-764
Burned Fireclay Super Scorifiers, HarbisonWalker
Crucibles
Versatile in service, Super Scorifiers may be used to separate precious metals and ores from incinerated scrap samples.
  • Manufactured from high quality fireclay
  • Pressed to consistent dimensions and burned under closely controlled conditions
  • Manufacturing process results in more heats per scorifier, and excellent thermal shock resistance