Sodium dodecyl sulfate (SDS) 10% in aqueous solution

Supplier: Rockland Immunochemical

MB-015 MB-015-1000
CAMB-015EA 182 CAD
CAMB-015 76236-394
Sodium dodecyl sulfate (SDS) 10% in aqueous solution
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MDL Number: MFCD00036175
CAS Number: 151-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR