Ammonium acetate 5 M in aqueous solution

Supplier: Rockland Immunochemical

Synonyms: Acetic acid ammonium salt

MB-041 MB-041
CAMB-041EA 182 CAD
CAMB-041 RLMB-041
Ammonium acetate 5 M in aqueous solution
Ammonium acetate
Formula: H₃CCOONH₄
MW: 77.08 g/mol
MDL Number: MFCD00013066
CAS Number: 631-61-8
https://ca.vwr.com/stibo/web/std.lang.all/06/04/16640604.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR