Corning® 1536-Well Cyclic Olefin Copolymer Microplates, Corning
Supplier: Corning
4370 4371 4372 4373 4675 4676 4677 4705
76175-630CS 3030.78 CAD
76175-630 76175-632 76175-634 76175-636 76175-638 76175-640 76175-642 76175-644
Corning® 1536-Well Cyclic Olefin Copolymer Microplates, Corning
Microplates HTS Microplates
1536-Well Cyclic Olefin Copolymer microplates are perfectly suited for high-throughput screening (HTS)
  • Bar coded on the short side of the microplate
  • Cyclic Olefin Copolymer (COC) material
  • Flat top for improved sealing
  • Ultra clear 127 µm film thickness
  • Variety of treatment options
  • Kalypsys and GNF system compatible
The ultraclear, 127 μm film bottom thickness is ideal for cell-based assays that require advanced optical analysis. The COC microplates are mechanically stable and inherently resistant to a broad range of solvents including DMSO and alcohols,More Product Information