Pierce™ Protein A/G and A/G Plus Agarose, Thermo Scientific
Supplier: Thermo Scientific
20421
20422
20423
CAPI20421EA
1212.27
CAD
CAPI20421
CAPI20422
CAPI20423
Pierce™ Protein A/G and A/G Plus Agarose, Thermo Scientific
Protein Purification
Protein Purification Resins
Pierce™ Protein A/G agarose and A/G Plus Agarose is a affinity resin for antibody purification.
- Immobilized Protein A/G is ideal for polyclonal IgG purification from nearly any mammalian species
- Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
- Protein A/G is immobilized by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
- Standard capacity beads with a normal load of immobilized Protein A/G