Pierce™ Recombinant Protein A Plus Agarose, Thermo Scientific
Supplier: Thermo Scientific
20365 20366
CAPI20365EA 1024.45 CAD
CAPI20365 CAPI20366
Pierce™ Recombinant Protein A Plus Agarose, Thermo Scientific
Protein Purification Protein Purification Resins
Pierce™ Recombinant Protein A agarose is a beaded agarose resin to which a genetically engineered form of Protein A has been immobilized for use in IgG purification applications where the native ligand has undesirable effects.
  • Immobilized Protein A is ideal for polyclonal IgG purification from human and rabbit
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Standard capacity beads with a normal load of immobilized Protein A
Although some non-essential regions have been removed, recombinant is similar to native Protein A in overall size and IgG-binding domains. The protein has four high affinity binding sites capable of specifically binding to the Fc region of immunoglobulin molecules from several species.