Pierce™ Protein A and A Plus UltraLink™ Resin, Thermo Scientific
Supplier: Thermo Scientific
53139
53142
CAPI53139EA
1309.03
CAD
CAPI53139
CAPI53142
Pierce™ Protein A and A Plus UltraLink™ Resin, Thermo Scientific
Protein Purification
Protein Purification Resins
Protein A UltraLink™ and Protein A Plus UltraLink™ resins are extremely durable beaded polyacrylamide supports to which IgG-binding Protein A has been immobilized for use in IgG purification methods
- Specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG
- Acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
Protein A resin is used to bind and purify IgG in gravity-flow, spin-column or medium pressure chromatography methods.