Bee-Safe® Hood, High-Tech Conversions
Supplier: High-Tech Conversions
GAH-HOOD-WH-L GAH-HOOD-BL-L
76169-758CS 718.58 CAD
76169-758 76169-760
Bee-Safe® Hood, High-Tech Conversions
Disposable Garments
This hood is a disposable, lightweight and economical solution with superior particulate filtration.
  • Spunbound polypropylene nonwoven
  • Disposable
  • Covers hair and neck
  • Lightweight
  • Superior particulate filtration
The Bee-Safe Hood provides hair and neck coverage that is lightweight for comfort and has superior particlate filtration for many different applications
Applications include cleanroom, food industry, medical, pharma, and safety. For use in laboratories and manufacturing environments.
SBPP nonwoven material. Latex-free elastic edges to minimize allergic reactions. Machine made to ensure consistency throughout production. Cleanroom packaging.
This hood is disposable and has very competitive pricing.
Packaging: 14 x 11 x 11"; 1000 Pieces/Case.