Stearic acid, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Octadecanoic acid, Octadecylic acid

569398-100GM 569398-25GM
CA80108-300EA 1188.84 CAD
CA80108-300 CA80108-302
Stearic acid, Sigma-Aldrich®
Stearic acid
Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
MDL Number: MFCD00002752
CAS Number: 57-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR