Flat Carbon Fiber Soft Tip, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78751-01 78751-02 78751-03 78751-04
103302-354EA 70.85 CAD
103302-354 103302-356 103302-358 103302-360
Flat Carbon Fiber Soft Tip, Electron Microscopy Sciences
Forceps
Tweezers with scratch resistant carbon fiber tips, specially designed to handle sensitive objects.
  • Stainless steel body
  • Corrosion resistant tips
  • ESD safe
  • Length 110 mm
  • Tip width 2×2×2, 2.5, 6, and 10 mm
The body of the EMS carbon fiber soft tip tweezers is made from low carbon austenitic, non-magnetic stainless steel
Typical applications include handling of sensitive and electronic components, soft materials, micro-mechanical parts, glass, quartz, ceramics and silicon wafers. The tweezers body is made from non-magnetic stainless steel. ESD safe.
The tips are corrosion resistant to moderately aggressive chemicals, salts and acids. Optimal working temperatures are up to 350 °C (662 °F). For short term use, maximum temperature should not exceed 925 °C (1697 °F).