Cupellation Muffle Furnace, 100-Place, Furnace and Assay Supplies

Supplier: KLEN INTERNATIONAL (74) PTY LTD.
FAS-170-NG1 FAS-170-OIL FAS-170-LPG1 FAS-170-NG2 FAS-170-LPG2
CA11023-212EA 116411.29 CAD
CA11023-212 CA11023-214 CA11023-216 CA76047-116 CA76047-118
Cupellation Muffle Furnace, 100-Place, Furnace and Assay Supplies
Ovens Furnaces (Ashing and Muffle Ovens)
Available with natural gas, liquefied petroleum gas, or oil (diesel) fuel source options

  • Suitable for labs 400-1000 samples per day including standards.
  • Base construction uses high-quality 75 x 75 RHS
  • Furnace body construction is durable 2mm laser-cut plate
  • High quality refractory brick and insulation
  • Bolt holes in base plates of all 4 legs for individual adjustment on uneven surfaces
  • Pneumatic door controls with hand valve—optional foot pedal available
  • Optional port viewing window in door
  • Vent tube height 1110mm

This furnace has a fan forced singleMore Product Information
Order Now

SPECIFICATIONS

Shipping dimensions 154 x 149 x 198 cm
Environmentally Preferable
Shipping weight 1080 kg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR