Large Double Slot Vise SEM Specimen Holder 25 mm, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75947-HH 75947-H
103302-148EA 132.25 CAD
103302-148 103302-450
Large Double Slot Vise SEM Specimen Holder 25 mm, Electron Microscopy Sciences
Specimen Mounting Supplies
Clamps thin specimens or cross sections without conductive paint or tape for scanning electron microscopy.
  • Made of machined aluminum with two stainless steel allen set screws
Large double-slot set screw vise scanning electron microscope specimen holder with (2) 2.5× 5 mm deep slots.
Ordering information: Includes allen wrench.