JEOL SEM Mount, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75702 75703 75704
103302-544EA 87.86 CAD
103302-544 103306-028 103306-204
JEOL SEM Mount, Electron Microscopy Sciences
Specimen Mounting Supplies
JEOL SEM mounts are made from lathe finished ultra-pure aluminum for scanning electron microscopy.
Head diameter: 11/4" (32 mm)
Height: 3/8" (10 mm)