Sodium acetate 3 M pH 5.2

Supplier: G-Biosciences

R010
CA82023-096EA 161.6 CAD
CA82023-096
Sodium acetate 3 M pH 5.2
Sodium acetate
Formula: H₃CCOONa
MDL Number: MFCD00012459
CAS Number: 127-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR