Cupellation Muffle Furnace, 50-Place , Furnace and Assay Supplies

Supplier: KLEN INTERNATIONAL (74) PTY LTD.
FAS-160-NG2 FAS-160-LPG2
CA76047-124EA 44266.92 CAD
CA76047-124 CA76047-126
Cupellation Muffle Furnace, 50-Place , Furnace and Assay Supplies
Ovens Furnaces (Ashing and Muffle Ovens)
Available with natural gas, liquefied petroleum gas, or oil (diesel) fuel source options

  • Suitable for small labs 100-400 samples per day including standards
  • Base construction uses high-quality 50 x 50 RHS
  • Furnace body construction is durable 2mm laser-cut plate
  • High quality refractory brick and insulation
  • Bolt holes in base plates of all 4 legs for individual adjustment on uneven surfaces
  • Counterbalanced door with manual lever to open
  • Optional port viewing window in door
  • Vent tube height 1160mm

This furnace has a fan forced single burner system with an automatic ignition system. Temperature is monitored and modulated automatically by a Type K thermocouple system and Omron temperature controller, pre-set for a maximum temperature of 1100 degrees Celsius. This controller is simple to operate, and has a 2-level display (PV and SV), that shows actual temperature as well as the temperature the furnace is set to.

Cupels are 50 x 6A, 50 x 7A or 7AS.

Ordering information: For oil model or optional equipment, contact your sales representative.

Caution: Please note that a minimum of 1 meter clearance around the furnace is required.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR