ZymoBIOMICS™ DNA Kits, Zymo Research
Supplier: Zymo Research
D4300 D4301 D4302 D4304 D4305 D4306 D4300T D4308
77001-494EA 704.9 CAD
77001-494 77001-504 77001-506 77001-524 77001-526 77001-528 76197-094 76211-516
ZymoBIOMICS™ DNA Kits, Zymo Research
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The ZymoBIOMICS™ DNA Kits are designed for purifying DNA from a variety of sample inputs that is immediately ready for microbiome or metagenomic analyses.
  • Ultra-pure, inhibitor-free DNA from many microbiomic sample types (feces, soil, water, biofilms, swabs, body fluid, etc.) that is ideal for all downstream applications including PCR, arrays, 16S rRNA gene sequencing, and shotgun sequencing
  • Innovative lysis system enables efficient and unbiased lysis of microbes including Gram-positive/negative bacteria, fungus, protozoans, and algae for accurate microbial community profiling
  • Provides superior yields when compared to other suppliers
The ZymoBIOMICS® DNA Kits are designed for purifying DNA from a variety of sample inputs that is immediately ready for microbiome or metagenome analyses. The ZymoBIOMICS® lysis system eliminates bias associated with unequal lysis efficiencies of different organisms (e.g. gram-negative/positive bacteria, fungus, protozoans, and algae) making it ideal for microbial community profiling. Uniform mechanical lysis of tough microbes is achieved by bead beating with the innovative ultra-high density BashingBeads™.This kit is equipped with our OneStep™ PCR Inhibitor removal technology, enabling PCR reaction from inhibitor-rich environmental samples. Purified DNA ideal for all downstream applications including PCR, arrays, 16S rRNA gene sequencing, and shotgun sequencing. DNA size is 15 to 20 kb.