Tweezers WA1 Handle with 22-ESD Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
TIP-22-ESD 10004-E22-SM
103301-850EA 19.64 CAD
103301-850 103302-568
Tweezers WA1 Handle with 22-ESD Tips, Electron Microscopy Sciences
Forceps
Swiss made precision tweezers from Dumont.
  • Hand made of the highest quality steels and alloys
  • Perfectly finished to suit your application
These tweezers have an anti-magnetic WA1 handle with replaceable 22-ESD tips. Tips attach with screws. Length is 125 mm.
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox.