HRP-Conjugate Diluent, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62716 62718-15 62717 62718
103304-006EA 393.01 CAD
103304-006 103305-870 103306-246 103307-080
HRP-Conjugate Diluent, Electron Microscopy Sciences
Buffers Assay Buffers
Antibody-diluent buffer with additional component for stabilizing HRP-conjugate.
  • For preparing solution of HRP-conjugate used as detection reagent
  • Antibody-dilutent buffer with additional component for stabilizing your HRP-conjugate
  • Can further standardize the assay preparing ready-to-use conjuigate solutions
Specifically designed for preparing solution of your HRP-conjugate used as the detection reagent. Allows user to further standardize the assay preparing ready-to-use conjugate solutions in advance and store them in refrigerator without loss of activity.