Tef Cap Industries Heat Exchange Tubing, Ace Glass
Supplier: Ace Glass
12067-15 12067-20 12067-25
76107-042FT 77.36 CAD
76107-042 76107-044 76107-046
Tef Cap Industries Heat Exchange Tubing, Ace Glass
Tubing Transfer Tubing
Heat exchange tubing made from copper, encapsulated in PTFE.
  • For use with heating and cooling coils on pilot plant reactors
The tubing also allows you to make your own custom heating or cooling exchanger.