Earth-Friendly Long Handle Sampling Spoons, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72133 72131 72132
103301-936EA 75.93 CAD
103301-936 103302-330 103302-340
Earth-Friendly Long Handle Sampling Spoons, Electron Microscopy Sciences
Spoons
Long handles ideal for acquiring samples from bottles, jars, and more.
  • Length 17.8 cm
  • Non-sterile
  • Translucent
  • Available in 3 bowl sizes
Long-handled spoons are ideal for acquiring samples from bottles, jars, and other confined areas and helping to avoid personal contact with sample materials. A flat edge on the rim of each bowl allows fast levelling to collect and measure small amounts with accuracy. Each spoon is 17.8 cm (7") overall length. Non-sterile and translucent in color.