Water, molecular grade

Supplier: G-Biosciences

Synonyms: Hydrogen oxide

786-72C 786-73C
CA82021-430EA 441.59 CAD
CA82021-430 CA82021-434
Water, molecular grade
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR