Water, DEPC treated

Supplier: G-Biosciences

786-109 786-117 786-118
CA82021-260EA 280.51 CAD
CA82021-260 CA82021-262 CA82021-264
Water, DEPC treated
Water

Ultra pure DEPC-Treated water is suitable for use with sensitive RNA research. It is prepared by incubating with 0.1% DEPC (diethylpyrocarbonate), an RNase inhibitor, and then autoclaved to remove the DEPC, to prevent downstream interference, and sterilized.

Formula: H₂O
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR