N-Ethyldiisopropylamine ≥99.5%, Biotechnology Grade, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Hünig’s base, N,N-Diisopropylethylamine, N.N’-Diisopropyl ethylamine, DIPEA, Ethyldiisopropylamine

496219-100ML 496219-2L
CABJ496219100MEA 157.94 CAD
CABJ496219100M CABJ496219-2L
N-Ethyldiisopropylamine ≥99.5%, Biotechnology Grade, Burdick & Jackson™
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
Boiling Pt: 128.33 °C
Melting Pt: –19.99 °C
Density: 0.75 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
EINECS: 230-392-0
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.45 %
Water (Karl Fischer) Max. 0.05 %
APHA Max. 20


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR