N,N-Dimethylformamide 99,9, Biotechnology Grade, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF

494488-2L 494488-6X1L 494488-1L
CABJ494488-2LEA 347.24 CAD
CABJ494488-2L CABJ4944886X1L CABJ494488-1L
N,N-Dimethylformamide 99,9, Biotechnology Grade, Burdick & Jackson™
N,N-Dimethylformamide

  • Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
  • As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
  • Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.


Beilstein Number: 605365

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153
Melting Pt: -61
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 57.5 °C (1.013.25 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3 III
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.005 %
Amines (as dimethylamine) Max. 8 ppm
Titrable Acid Max. 0.0005 meq/g
Titrable Base Max. 0.003 meq/g
Absorbance at 400 nm Max. 0.01
Absorbance at 340 nm Max. 0.01
Absorbance at 310 nm Max. 0.05
Absorbance at 295 nm Max. 0.10
Absorbance at 275 nm Max. 0.30
Absorbance at 270 nm Max. 1.00


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR