Dichloromethane ≥99.9% stabilized, Biotechnology Grade, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Warning


Stabilization: Stabilised with amylene 40-150 ppm

494453-1L 494453-2L
CABJ494453-1LEA 165.11 CAD
CABJ494453-1L CABJ494453-2L
Dichloromethane ≥99.9% stabilized, Biotechnology Grade, Burdick & Jackson™
Dichloromethane
Formula: CH₂Cl₂
Boiling Pt: 39…41 °C (1.013 hPa)
Melting Pt: –97 °C
Density: 1.32…1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.85 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.002 %
2-methyl-2-buten (GC) 40 - 150 ppm
Absorbance at 400 nm Max. 0.01
Absorbance at 340 nm Max. 0.01
Absorbance at 260 nm Max. 0.02
Absorbance at 250 nm Max. 0.05
Absorbance at 240 nm Max. 0.20
Absorbance at 235 nm Max. 1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR