Methanol ≥99.93%, BioReagent for protein sequencing, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

494437-1L 494437-2L
CABJ494437-1LEA 145.45 CAD
CABJ494437-1L CABJ494437-2L
Methanol ≥99.93%, BioReagent for protein sequencing, Burdick & Jackson™
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6
Melting Pt: -98
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 II
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.925 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.02 %
Titrable Acid Max. 0.0003 meq/g
Titrable Base Max. 0.0002 meq/g
Absorbance at 400 nm Max. 0.01
Absorbance at 260 nm Max. 0.01
Absorbance at 240 nm Max. 0.05
Absorbance at 230 nm Max. 0.15
Absorbance at 220 nm Max. 0.30
Absorbance at 210 nm Max. 0.60
Absorbance at 205 nm Max. 1.00
Application test complying
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR