[Lys(Me3)27]-Histone H3 (21-44)-GK,Biotin

Supplier: Anaspec

AS-64367-025 AS-64367-1
103008-008EA 309.23 CAD
103008-008 103008-010
[Lys(Me3)27]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides

This peptide is Histone H3 amino acid residues 21 to 44 tri-methylated at Lys-27 with an addidtional C-terminal glycine followed by a biotinylated lysine. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ATKAAR-K(Me3)-SAPATGGVKKPHRYRPG-GK(Biotin)
MW:2959.5 Da
% peak area by HPLC:95
Storage condition:-20° C

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR