[Lys(Me2)27]-Histone H3 (21-44)-GK(Biotin),Biotin
Supplier: Anaspec
AS-64366-025
AS-64366-1
103008-004EA
309.23
CAD
103008-004
103008-006
[Lys(Me2)27]-Histone H3 (21-44)-GK(Biotin),Biotin
Proteins and Peptides
This peptide is Histone H3 amino acid residues 21 to 44 di-methylated at Lys-27 with an additional C-terminal glycine followed by a biotinylated lysine. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ATKAAR-K(Me2)-SAPATGGVKKPHRYRPG-GK(Biotin)
MW:2945.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...