[Lys(Me2)9]-Histone H3 (1-21)-GGK,Biotin
Supplier: Anaspec
AS-64359-025
AS-64359-1
103007-976EA
309.23
CAD
103007-976
103007-978
[Lys(Me2)9]-Histone H3 (1-21)-GGK,Biotin
Proteins and Peptides
This peptide is Histone H3 amino acid residues 1 to 21 di-methylated at Lys-9 with a C-terminal GG linker followed by a biotinylated lysine. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ARTKQTAR-K(Me2)-STGGKAPRKQLA-GGK(Biotin)
MW:2751.2 Da
% peak area by HPLC:95
Storage condition:-20° C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...