Quality Economy Brand Luer Slip Syringes with Attached Needle, Air-Tite
Supplier: Air-Tite
MS12558 MS12712 MS3181 MS3201 MS32234 MS3221 MS32558 MS5181
76124-672BX 27.04 CAD
76124-672 76124-674 76124-676 76124-678 76124-680 76124-682 76124-684 76124-686
Quality Economy Brand Luer Slip Syringes with Attached Needle, Air-Tite
Syringes General-Purpose Syringes
Quality disposable syringes with needle, made to exacting specifications. Great value at a low cost.
  • Standard 3-part syringe
  • Latex-free
  • Non-toxic
  • Pyrogen-free
  • Positive plunger stop
Why spend more if you are looking for a basic, functional syringe? The Air-Tite brand syringes are a quality syringe, manufactured under ISO standards. Suitable for laboratory and veterinary use, these basic 3-part syringes offer a lubricated rubber tip plunger for smooth gliding in sterile, individually wrapped packaging.
From 1 to 5 ml in luer slip with attached needle,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable