VWR® Digital Handheld Refractometers
Supplier: VWR International
38-922 38-923
75997-572EA 699.86 CAD
75997-572 75997-574
VWR® Digital Handheld Refractometers
Refractometers Handheld Refractometers
Dual scale handheld refractometers designed to provide ranges of Brix (sugar concentration) and refractive index measurements.
  • Optical glass prism and stainless steel dish to stabilize sample temperature
  • Simple zero calibration with distilled water
  • Automatic temperature compensation (ATC) to correct readings of water and sucrose solutions to 20°C
Ordering information: Supplied with two batteries and a battery indicator, multi-language guide and protective case.