LiChroCART® Frit Elements, MilliporeSigma
Supplier: MilliporeSigma
LiChroCART®
1.51496.0001 1.51496.0001
CA1.51496.0001CS 77282.57 CAD
10143-992 CA1.51496.0001
LiChroCART® Frit Elements, MilliporeSigma
Chromatography Supplies Chromatography Spare Parts
These sealing elements are suitable for 4.6 and 4 mm cartridges LiChroCART®.