Human;Pig Endothelin 1
Supplier: Anaspec
AS-22861
AS-22859
AS-22860
102996-308EA
760
CAD
102996-308
102999-368
102999-370
Human;Pig Endothelin 1
Proteins and Peptides
ET-1 is a potent vasoconstrictor peptide derived from endothelial cells. It plays a role in regulation of cardiovascular functions
Sequence:CSCSSLMDKECVYFCHLDIIW (Disulfide bridge: 1-15 and 3-11)
MW:2492 Da
%Peak area by HPLC:≥95%
Storage condition: -20°C
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...