Antibody Diluent, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62713 62713-01 62714 62714-01 62715 62715-01
103305-764EA 412.66 CAD
103305-764 103306-158 103306-248 103306-262 103307-090 103307-116
Antibody Diluent, Electron Microscopy Sciences
Buffers Antibody Purification Buffers
These can be used as buffer for diluting primary and secondary antibodies.
  • Reduces non specific binding of the antibodies
  • Minimizes negative effects of disturbing substances
  • Minimizes low or medium affinity cross-reactivities of the antibodies
  • More reliable results
These can be used for diluting antibodies if they were stored even at –20 °C in glycerol, or in refrigerator
For 'trouble-giving' antibodies and in situ PCR applications, this dilutent may be used as a washing buffer, preventing secondary binding of analystes during washing.
When used in pathology, it also greatly reduces non-specific reactivity of human serum components and immunoglobulins in tissue, vessels and cells with mouse antibodies used on section. Excellent for IHC, flow cytometry on fixed cells, western blot and other immune assays.
Order Now
SPECIFICATIONS
Environmentally Preferable
Storage temperature 4 - 8 °C