Pierce™ Glycoprotein Isolation Kit, ConA and WGA, Thermo Scientific
Supplier: Thermo Scientific
The Glycoprotein Isolation Kit allows isolation of glycoproteins from complex mixtures, including serum, tissue and cultured cell lysates.
- Higher glycoprotein recovery when compared to other kits
- Glycoprotein purification in less than an hour
- Lectin does not leach from resin when processing sample
- Dialysis or protein precipitation of recovered glycoproteins is not required prior to protein assay
The procedure uses concanavalin A (ConA) or wheat germ agglutinin (WGA) as a ligand to bind a specific class of glycoprotein from complex protein samples - those containing alpha-lined mannose or terminal glucose residues (in case of ConA) and N-Acetyl glucosamine (GlcNAc) groups or sialic acid (in case of WGA). This kit has been used to isolate glycoproteins from serum as well as HeLa and CHO cell lysates in approximately 50 minutes.
Delivery information: Kit contains ConA lectin resin (or WGA lectin resin) (1.1 mL), Binding/wash buffer (5X), (6.5 mL), Elution buffer (5 mL), Spin columns (10) and collection tubes (20).