Polyethylenimine hydrochloride, M.W. 160000 PEI MAX, linear

Warning

75800-190EA 686.65 CAD
75800-190
Polyethylenimine hydrochloride, M.W. 160000 PEI MAX, linear
Polyethylenimine hydrochloride

Linear polyethylenimine HCl is a fully hydrolyzed (deacylated) and highly water soluble.


  • Molecular Weight: 160,000 (~87,000 free base)


PEI MAX 160000 is supplied as a hydrochloride salt for ease of handling. It can be converted into the linear free amine (PEI) form by neutralization with base.


Linear polyethylenimine is a highly-charged cationic polymer that readily binds highly anionic substrates. Industrially these polymeric amines are useful in many applications derived primarily from its polycationic character that allows it to tightly bind to negatively charged substrates.

Formula: (C₂H₅N)n·HCl
Storage Temperature: Ambient
CAS Number: 9002-98-6

Order Now

Specification Test Results

Appearance White to off-white solid
Solubility Soluble in cold and room temperature water, Insoluble in common organic solvents (ethanol, acetone, tetrahydrofuran)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR