GL45 Cap with NPT Tap, Ace Glass Incorporated
Supplier: Ace Glass
12703-11 12703-12 12703-13 12703-14 12703-21 12703-22 12703-23 12703-24
76107-058EA 226.34 CAD
76107-058 76107-060 76107-062 76107-064 76107-066 76107-068 76107-078 76107-080
GL45 Cap with NPT Tap, Ace Glass Incorporated
Closures Bottle Closures
GL45 cap with an NPT tap designed to accept various male NPT adapters found, for instance, in ACE compression tube fittings or standard taper joint adapters.
  • Caps are made of either polypropylene or PTFE
  • Ideal connections for Duran laboratory bottles
  • Caps accept various male NPT adapters
Order Now
SPECIFICATIONS
Environmentally Preferable