PTFE Bottom Surface Cover for Stainless Steel Flat Head, Ace Glass Incorporated
Supplier: Ace Glass
6467-13 6467-14 6467-11 6467-12
76107-074EA 707.57 CAD
76107-074 76111-088 76107-070 76107-072
PTFE Bottom Surface Cover for Stainless Steel Flat Head, Ace Glass Incorporated
Lids Reaction Vessel Lids
PTFE bottom surface cover will protect stainless steel flat heads from chemical attack.
  • Cover made of chemical-resistant PTFE
  • Secured by the coupling and the joint inserts
  • For use with KF flat flanged stainless steel heads